Light Pipe Scintillators, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
82001-100 82000-89 82000-74 82000-104 82001-88 82000-X 82001-120 82000-10 82001-X 82008-RS 82004-60 82005-60
100495-410EA 1556.93 CAD
100495-420 100495-410 100495-422 100495-412 102099-374 100502-322 100495-418 100502-324 100495-424 100495-414 100502-320 100495-416
Light Pipe Scintillators, Electron Microscopy Sciences
Lights Microscope Bulbs
The light pipe is the connection between the scintillator and the phosphomultiplier tube (PMT). It transfers the photons produced by the scintillator for detection by the PMT.
  • Scintillator material coated directly on the light pipe which improves the signal
  • Re-coating is typically needed twice per year
Light Pipes are available in optical quality quartz and optical grade acrylic. Cambridge reduced-tip models are only available in acrylic.
Delivery information: Includes one Re-coating Certificate. High voltage wire and corona ring are included, JEOL pipes include a flange.