Bright-Line Counting Chamber, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
63514-32 63514-34 63514-11 63514-30 63514-12 63514-20 63514-31 63514-10
100498-508EA 169.57 CAD
100498-508 100498-510 100503-092 100498-504 100498-502 100502-518 100498-506 102094-786
Bright-Line Counting Chamber, Electron Microscopy Sciences
Slides Counting Chambers
For accurate counting of blood cells platelets, dust/yeast cluster and specimen samples. Sharp contrast from rhodium plated on "V-Load". Triple dividing lines clearly delineate counting area for accurate and precise results. Molded from a single piece of thermal and shock resistant glass with a polished chamber and cover glass assuring precise capillary loading and even cell distribution. Fluid volume is 0.1 mm3/mm2 and 0.00025 mm2 per each square.
Delivery information: The complete set includes one counting chamber, two 0.4 mm cover glass, one red cell pipette, one white cell pipette, tubing, mouthpieces, and storage case.