Large Glass Slides, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71862-01 71860-01
100501-330EA 212.43 CAD
100501-330 100491-372
Large Glass Slides, Electron Microscopy Sciences
Slides Microscope Slides
These slides are precleaned, individually selected and are suitable for large specimen sections, such as, brain tissue.
In addition to our comprehensive collection of glass slides manufactured by ESCO we now carry large glass slides which are available in 75×38 mm and 75×51 mm.