Large Gelatin Capsules, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70116 70115 70118 70114 70117
100500-586BX 55.05 CAD
100500-586 100500-588 100500-590 100500-592 100500-594
Large Gelatin Capsules, Electron Microscopy Sciences
Specimen Mounting Supplies
Our large size gelatin capsules are ideal for storing either delicate or small items. It is also suitable for use as an embedding mold for paraffin or paraplast. The capsules come complete with a locking ring. A variety of sizes are available.