Lacey Carbon Film, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
LC305-CU LC200-NI-25 LC200-CU-25 LC305-NI LC200-AU LC200-AU-25 LC200-CU LC200-NI LC300-AU-25 LC325-AU-25 LC325-CU-25 LC325-NI-25 LC400-AU-25 LC400-CU LC400-CU-25 LC400-NI LC400-NI-25 LC200-CU-100 LC200-CU-150 LC300-CU-100 LC300-CU-150 LC400-CU-100 LC400-CU-150 LC200-NI-100 LC200-NI-150 LC300-NI-100 LC300-NI-150 LC400-NI-100 LC400-NI-150 LC200-AU-100 LC200-AU-150 LC300-AU-100 LC300-AU-150 LC400-AU-100 LC400-AU-150 LC200-AU-UL LC200-CU-UL LC200-NI-UL LC300-AU-UL LC300-CU-UL LC300-NI-UL LC400-AU-UL LC400-CU-UL
100491-192EA 58.51 CAD
100491-192 100491-188 100491-184 100491-194 102101-424 102101-426 102101-428 102101-430 102101-434 102101-436 102101-438 102101-440 102101-444 102101-446 102101-448 102101-450 102101-452 103302-712 103302-714 103302-716 103302-750 103302-752 103302-754 103302-890 103302-892 103302-894 103302-896 103302-898 103302-900 103305-112 103305-114 103305-116 103305-118 103305-120 103305-122 76439-834 76439-848 76439-862 76439-870 76439-878 76439-880 76439-918 76439-932
Lacey Carbon Film, Electron Microscopy Sciences
Graticules, Reticles and Grids
Lacy carbon film provides support without interfering with transmission electron microscopy observation.
  • Copper, nickel and gold grids in 200, 300, and 400 mesh grids
  • Optically checked followed by batch testing in the EM
This carbon coated film on a broken pattern consists of woven-mesh-like holes of different sizes and shapes.
Ordering information: Molybdenum grids available on request.
Packaging: Packed in grid storage box.