Lab-Tek® II Chamber Slide™ System, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70360-26 70360-24 70360-46 70379-16 70360-12 70360-82 70379-44 70379-84 70379-46 70360-22 70379-86 70379-42 70360-14 70379-22 70379-26 70360-42 70360-44 70379-12 70379-82
100489-082EA 4290.27 CAD
100489-082 100489-080 100489-088 100489-118 100489-072 100489-090 100489-126 100489-130 100500-084 100489-078 100500-086 100489-124 100489-074 100489-120 100501-726 100489-084 100489-086 100489-114 100489-128
Lab-Tek® II Chamber Slide™ System, Electron Microscopy Sciences
Slides Chamber Slides
Lab-Tek® II Chamber Slide™ products offer optimal surface for excellent attachment and growth of cells, easily distinguished by the blue mark
  • Sterile, Chamber Slide™ products have removable, polystyrene media chamber of 1, 2, 4, and 8-well configuration
  • Low fluorescent glass microscope slide (25 x 75 x 1.2 mm) with rounded corners
  • Biocompatible adhesive
  • Polystyrene cover
  • Inert hydrophobic well border printed on slide
  • SuperFrost printed area
Lab-Tek® II CC2™ Chamber Slide™ products are the newest addition to our Lab-Tek® II Chamber Slide™ family. CC2 shares several similarities with the original Lab-Tek® II Chamber Slide, however, it’s pronounced difference is that the glass growth surface has been chemically modified. This modification provides an optimal surface for growing fastidious cells such as primary neurons or poorly adherent cell lines. CC2 also are non-auto fluorescent making them ideal for fluorescent applications such as FISH (Fluorescent in situ Hybridization). The growth surface remains stable without refrigeration.
Lab-Tek® II and Lab-Tek II CC2™ Chamber Slide™ Systems all come packed in a sterile sealed tray. 8 units per tray, 2 trays per pack and 12 trays per case.
This design allows for the removal of the media chamber and adhesive from the slide, using a separation device. The blue hydrophobic border is raised slightly and isolates the individual wells, and prevents cross contamination. The slide can be coversliped.