Lab-Tek® Chamber Slide™ System, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70421 70417 70420 70423 70384 70393 70407 70415 70418 70400 70381 70383 70406 70416 70410 70386 70419 70397 70396 70401 70422 70390 70394 70413 70380 70391 70411
100488-396EA 1679.63 CAD
100488-396 100500-616 100500-618 100500-620 100488-386 100488-390 100500-608 100500-614 100501-174 100501-766 100488-382 100488-384 100499-898 100499-900 100501-768 100499-894 100499-902 100501-764 100499-896 100500-604 100501-722 100500-598 100500-602 100488-394 100500-596 100500-600 100500-610
Lab-Tek® Chamber Slide™ System, Electron Microscopy Sciences
Slides Chamber Slides
Permanox® is a special molding resin, which meets the growing requirements of most anchorage dependent cells; also excellent chemical resistance allows for cell fixation and the staining of organic solvents for EM and other techniques. Low oxygen permeation makes Permanox® ideal for anoxic cell growth.
  • Developed for microscopy of in situ cell culture
  • Cells grow on a standard microscope slide
  • Media chamber can be removed when culturingis complete
  • Useful for viral and mycoplasma testing, chromosome studies toxicity tests and immunocytology
  • Permanox® plastic and glass do not interfere with fluorescent labels
The chamber slides combine a media-containing chambered super-structure with a standard soda lime glass slide or Permanox® plastic slide, sealing the two together with a biologically inert silicone gasket. After cells are grown, the chamber and gasket are easily removed. The slide may then be fixed, stained, cover slipped, and filed with the usual convenience. These slides, once fixed and stained, can be embedded using our Slide Duplicating Mold (Cat. 70170). Lab-Tek® Chamber Slide™ System all come packed in a sterile sealed tray, 8 units per tray, 2 trays per pack and 12 trays per case.