Kodak T-MAX 100 Professional Films, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
74160 74172 74164 74162 74157 74158
100488-102EA 793.76 CAD
100488-102 100500-692 100488-106 100488-104 102091-272 102091-274
Kodak T-MAX 100 Professional Films, Electron Microscopy Sciences
Photographic Film
Very high degree of enlargement, expanded exposure latitude.
  • Medium speed (EI-100)
  • Extremely high sharpness, fine grain
  • Total darkness
Especially useful for detailed subjects when maximum image quality is needed. Recommended for copying black and white photographs.
Recommended kodak developers: T-Max, D-76, HC-110, Microdol-X, Duraflo RT.