Biopsy Bags, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
62326-01 62326-05 62326-02 62326-15 62326-10
100490-170EA 34.14 CAD
100490-170 100501-992 103301-906 103302-090 103302-550
Biopsy Bags, Electron Microscopy Sciences
Bags Sample Bags
Bags are made up of porous paper which provides maximum tissue safety during processing.
  • Made from porous paper
  • Permits proper fluid exchange and drainage
  • Available in 25 and 35 mm diameter or 25 and 32 mm sizes
They permit proper fluid exchange and drainage.
To use: simply introduce the specimen into the bag and fold it enclosing the cassette or capsule. Now ready to process as usual.
Order Now
SPECIFICATIONS
Environmentally Preferable