Premium Abrasive Cut-off Wheels for Sectioning, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
50250-05 50250-09 50255-09 50250-03 50250-06 50250-07 50255-07 50255-10 50250-08 50255-03 50250-10 50255-04
100500-916BX 113.33 CAD
100500-916 100502-648 100500-938 100500-912 100500-918 100500-920 100500-934 100500-940 100500-922 100500-926 100502-650 100500-928
Premium Abrasive Cut-off Wheels for Sectioning, Electron Microscopy Sciences
Abrasives
Silicone Carbide disc are the finest wet or dry abrasive discs for sample preparation
Wheels are available in 9 different grit sizes, ranging from coarse to fine, on plain back or water resistant pressure sensitive adhesive back.
Pressure sensitive adhesive backed discs eliminates the need to transfer to plates. Silicon Carbide Abrasive Cut-Off Wheels offered long life, and affordably priced sectioning. All wheels are 9" (228.6 mm) diameter 0.062” (1.6 mm) thick; 1¼” (31mm) arbor.
Ordering information: Available in 8" diameter discs as a standard and 12" discs available upon request.