Paterson Safelight, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
74327-16 74327-10
100493-992EA 26.31 CAD
100493-992 100502-256
Paterson Safelight, Electron Microscopy Sciences
Lights Office Lamps
Perfect for the small darkroom, or for safety spot lighting individual work areas.
  • Freestanding, mounted on the wall, or ceiling
  • safe for all black-and-white papers
  • 120 V AC; 3-prong cord
DxH: 4x6.25"
Ordering information: The unit comes with a 15 W bulb.