White Translucent Cutting Mat, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
77621-18 77621-24 77621-12
100494-398EA 138.5 CAD
100494-398 100494-396 100501-484
White Translucent Cutting Mat, Electron Microscopy Sciences
Dissecting Boards
Cutting Mats will protect all table-top surfaces from knife cuts. Its smooth surface permits straight, even cutting and the cut mark heals instantly. Each mat is printed with both 1/2" and 1" grid patterns to aid in measuring and aligning work. Available in translucent white, which allows for cutting and trimming on a light box surface.