Ilford Multigrade IV RC Deluxe Photographic Papers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
83020-25 83020-10
101381-618EA 290.48 CAD
101381-618 101381-616
Ilford Multigrade IV RC Deluxe Photographic Papers, Electron Microscopy Sciences
Film Processing Supplies
This is a direct replacement for the Kodak Polycontrast IV RC B&W Papers. Neutral tone – Variable contrast. MULTIGRADE IV RC Deluxe has the standard weight of (190g/m2)with a resin coated base.
  • Resin Coated – Black & White
  • Non-Developer Incorporated
  • 12 Contrast Grades with MULTIGRADE Fibers
  • Glossy Surface
  • Compatible with the Kodak OC Filter
  • Dish/Tray Processing
  • Developer: Mutigrade, Bromophen or PQ Universal
  • Fixer: Ilford Rapid Fixer or Hypam
Order Now
SPECIFICATIONS
Environmentally Preferable