Nitrogen Purge Cabinet, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71219-20 71219-10 71219-30 71219-40
100493-150EA 2345.3 CAD
100493-150 100493-148 100493-152 100501-416
Nitrogen Purge Cabinet, Electron Microscopy Sciences
Desiccators Desiccator Cabinets
Acrylic, ethylene propylene, and Type 316 stainless steel welded parts
Simple and easy to use at temperatures up to 120°F. They can be used in freezers, refrigerators, or at room temperature. Cabinets have airtight doors that need no lubrication. Sliding perforated shelves for easy access, transparent acrylic wall for see-through storage. These cabinets are also available with Flow Meters. Direct reading scale flow meters with control levels that let you not only measure flow through the meter, but also make flow adjustments in the line.
Acrylic, ethylene propylene, and Type 316 stainlessMore Product Information