SECADOR™ 4.0 Desiccator Large Cabinet - Vertical, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71227-17 71227-16 71227-15 71227-20
100493-194EA 3706.65 CAD
100493-194 100493-192 100493-188 100493-198 100493-186 100493-190 100493-196
SECADOR™ 4.0 Desiccator Large Cabinet - Vertical, Electron Microscopy Sciences
Desiccators Desiccator Cabinets
SECADOR™ 4.0 Autodesiccator Cabinet - Vertical is a hygrometer with autodesiccator module which requires constant low humidity
The module contains 3 shelves, 4 positions and 2 latches. It resuces RH from 75% to less than 25% in 8 hours. Totally automatic, continual desiccation with silica beads, and generation every 20 minutes, keeps RH level in the SECADOR™ 4.0 Desiccator at a constant level. Two fans guarantee continuous airflow within the cabinet, and continuous exhaust of humid air.