Disposable Sterile Collecting Bottles, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64244-10 64244-02 64244-05
100490-500EA 746.74 CAD
100490-500 100490-496 100490-498
Disposable Sterile Collecting Bottles, Electron Microscopy Sciences
Bottles Cell Culture Bottles
Disposable polystyrene bottles for the storage of media, buffers and other aqueous solutions.
  • Easy grip sides facilitate handling
  • Sterile
  • Certified non-pyrogenic
  • Neck size 45 mm
Plug seal cap provides an airtight seal and helps minimize the risk of contamination.