Disposable Sterile Vacuum Filter Units, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
67045-12 67048-14 67045-11 67046-15 67047-11 67047-12 67047-14 67048-13 67048-16 67045-13 67048-15 67047-13 67047-15 67046-12 67046-11 67046-13 67048-12
100490-674EA 423 CAD
100490-674 100490-700 100490-672 100490-684 100490-686 100490-688 100490-692 100490-698 100490-704 100490-676 100490-702 100490-690 100490-694 100490-680 100490-678 100490-682 100490-696
Disposable Sterile Vacuum Filter Units, Electron Microscopy Sciences
Filters Bottle Top Filters
Ideal for filtration of tissue culture media, biological fluids, fixation buffers and more. (not for parenteral use.)
  • Vacuum filtration units fit standard threaded bottle tops
  • Designed for use on glass or polystyrene media bottles
  • Attaches to most popular 33 and 45 mm Ø bottle's necks with standard GPI threads
Units are color coded by membrane type (CA: Orange; NYL: Red, CN: Blue; and PES: Yellow). Choice of membrane media: Cellulose acetate (CA), for low protein binding. Nylon (NYL), for low extractable. Cellulose Nitrate (CN), for general purposeMore Product Information