Veco Square Mesh Oyster Grids, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
D75-CU D1002D-CU D100B-NI D100B-CU D75-NI
100495-514EA 97.36 CAD
100495-514 100495-490 100495-496 100495-494 100495-520
Veco Square Mesh Oyster Grids, Electron Microscopy Sciences
Graticules, Reticles and Grids
Veco grids offer superior handling characteristics.
Veco grids are the most rigid grids available. Measures 3.05 mm in diameter, with a 0.8 mil thickness. Material: Copper (Cu); Nickel (Ni); Gold (Au).