Veco Square Mesh Handle Grids, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
HD200-NI HD400-NI HD100-NI HD100S-NI HD100S-CU HD400-CU HD300-CU HD300-NI HD200-CU HD150-CU HD111S-CU
100495-854EA 61.02 CAD
100495-854 100495-870 100495-838 100495-840 100502-348 100501-562 100501-560 100495-864 100501-558 100501-556 100502-350
Veco Square Mesh Handle Grids, Electron Microscopy Sciences
Graticules, Reticles and Grids
Veco grids offer superior handling characteristics.
Veco grids are the most rigid grids available. Measures 3.05 mm in diameter, with a 0.8 mil thickness. Material: Copper (Cu); Nickel (Ni); Gold (Au).