Embra Square Mesh, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
E100-SS E100-TI E300-MO E100-AL E200-AL E300-AL E300-TI E200-MO E200-SS
100495-538EA 130.79 CAD
100495-538 100495-540 100495-562 100495-534 100495-542 100495-560 100495-564 100495-544 100495-548
Embra Square Mesh, Electron Microscopy Sciences
Graticules, Reticles and Grids
EMBRA electroformed grids combine a high open area with a rigid construction which allows for relatively easy handling. Measures 3.05 mm in diameter and 16 micron in thickness. Material: Copper (Cu), Nickel (Ni), Gold (Au), Stainless Steel (SS), Titanium (Ti), Molybdenum (Mo), Aluminum (Al).