Square and Grid Scales, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
68011-19 68011-16 68015-16 68015-19 68015-21 68018-16 68018-21 68018-19 68011-21
100499-148EA 319.71 CAD
100499-148 100499-144 100499-170 100499-172 100499-174 100499-188 100499-196 100499-192 100499-150
Square and Grid Scales, Electron Microscopy Sciences
Graticules, Reticles and Grids
Simple grids are convenient for making sketches of the obseved speimen on graph paper.
They are also useful for particle counting.