Rotatrim® Mastercut Quality Rolling Trimmers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
74880-17 74880-36 74881-16 74880-15 74880-30 74881-26
100494-160EA 1375.93 CAD
100494-160 100494-166 100494-168 100494-158 100494-164 100494-170
Rotatrim® Mastercut Quality Rolling Trimmers, Electron Microscopy Sciences
Cutters
Exclusively from Rotatrim®, its double steel tube suspension guides the cutting head with the stability essential for accurate, repeatable cuts.
Trimmers are for precision cutting. It can slice through tough materials like cardboard, litho plates, mount board, and even 1/16" hardboard; yet, Rotatrim® also has a delicate touch, trimming film, paper, and dry mount tissue to hairline tolerances. Known for its safe operation and best performance, the rotary blade is Sheffield tungsten steel, self-sharpening, and is completely shielded by a protective housing. Its blade assures consistentMore Product Information