Gilder Revolutionary Specimen Support Grids, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
T1500-NI T2000-CU T601H-NI T2000-NI T601-CU T1500-CU T601H-CU T601-NI
100502-356EA 188.18 CAD
100502-356 100496-030 100501-570 100502-358 100496-062 100496-028 100496-064 100501-572
Gilder Revolutionary Specimen Support Grids, Electron Microscopy Sciences
Graticules, Reticles and Grids
Type Hexagonal and Square represent our efforts to reduce the grid bar width (only 5 microns) enabling more of the specimen to be viewed
In addition to our square and hexagonal mesh Gilder Thin Bar Grids, we are now able to produce a very fine mesh that values up to 2,000 lines/inch. There is an increasing need in TEM for support thin films, routinely carbon, as thin as 1.5 - 2.0 nm. The pitch (the distance from the center of one bar to the center of the next bar) dimension in all grids remains constant, which allows them to be used as a lower magnification calibration aid.
Order Now
SPECIFICATIONS
Environmentally Preferable