Reusable/ Disposable Desiccant Cartridge, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71212-04 71212-02
100489-256EA 277.04 CAD
100489-256 100489-254
Reusable/ Disposable Desiccant Cartridge, Electron Microscopy Sciences
Desiccators Desiccants
A highly porous membrane covers the dish of the desiccant providing good moisture absorption.
These desiccant cartridges can be used in desiccator cabinets or non-vacuum desiccators, analytical balance cases, and precision laboratory instruments. The clear polystyrene dish allows for instant visual inspection of the silica gel color which changes from blue to pale pink as it absorbs moisture.
A highly porous membrane covers the dish of the desiccant providing good moisture absorption. The desiccant can be regenerated by heating cartridges in an oven at 50°C (122°F) overnight. For rapid
More Product Information