Replacement Desiccator Plates, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71242-23 71242-15
100493-224EA 445.56 CAD
100493-224 100501-418 100493-222
Replacement Desiccator Plates, Electron Microscopy Sciences
Desiccators Desiccators, Plates
These 4.8 mm thick plates are able to withstand temperatures up to 150°C (300°F) and have excellent resistance to thermal shock, and to chemicals. They are non-combustible and asbestos free. Round plates are perforated with 4 mm (0.165”) diameter holes on 12.7 mm (1/2”) centers in a straight line pattern. They are available in three sizes, and fit our desiccators.