Micro Cleaning Solution, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71054 71057 71055
100488-218EA 74.1 CAD
100488-218 100496-314 100496-312
Micro Cleaning Solution, Electron Microscopy Sciences
Cleaning Agents Detergents and Surface Cleansers
Specially designed for ultrasonic cleaning machines. Cleans glass, most metals, plastics, and rubber.These can work with any concentration.
  • Biodegradable
  • Contains no phosphates
  • Removes radioactive contaminants
Not to be used on zinc, magnesium, or aluminum.