Kempad-Pan W Polishing Cloths, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
50310-15 50310-05
100489-998EA 138.16 CAD
100489-998 100489-996
Kempad-Pan W Polishing Cloths, Electron Microscopy Sciences
Wipes Cleaning Kits and Pads
A non-woven textile cloth which is both oil and water resistant.
  • Excellent flatness
Recommended for intermediate and final polishing with diamond abrasive compounds. Available with Pressure Sensitive Adhesive (PSA) with and without hole.