Instrument Trays With Cover, Natural polypropylene, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
63312-01 63312-03 63312-02
100498-404EA 114.11 CAD
100498-404 100498-408 100498-406
Instrument Trays With Cover, Natural polypropylene, Electron Microscopy Sciences
Trays/Dishes/Pots Instrument Trays/Dishes
The cover protects the instruments from contamination.
  • Natural polypropylene
  • They are easily cleaned and sterilized
Steam autoclavable at 121 °C (250 °F), smooth with rounded corners.