Instrument Trays Blue polypropylene, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
63310-02 63310-06
100498-400EA 93.46 CAD
100498-400 100498-402
Instrument Trays Blue polypropylene, Electron Microscopy Sciences
Trays/Dishes/Pots Instrument Trays/Dishes
Lightweight blue polyethylene tray with smooth surface and rounded internal corners.
  • These deep trays keep fluids from splashing
  • Easy to clean
  • It is lightweight and noiseless as compared to heavy metal trays
  • Autoclavable at 275 °F (135 °C)
  • Available in 2 sizes