Inoculating Loops, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
62437-02 62436-03 62433-05 62437-03 62434-03 62434-02 62437-05 62436-05
100498-104EA 405.02 CAD
100498-104 100498-100 100498-086 100498-106 100498-084 100498-090 100498-088 100498-108 100498-102
Inoculating Loops, Electron Microscopy Sciences
Spreaders Inoculating Loops and Needles
Our high quality inoculating loops are made from platinum wire. The 15% iridium adds necessary rigidity permitting thinner wire for delicate work. The loop section is welded and smoothly finished to form a “perfect circle”.