Inoculating Loop and Needle Holders, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
62440-08 62440-06
100498-116EA 382.38 CAD
100498-116 100498-114
Inoculating Loop and Needle Holders, Electron Microscopy Sciences
Dissecting and Inoculating Loop and Needle Holders
The innoculating loops are made from durable, heat resistant brass with a fiber glass insulating sleeve over the gripping area. The threaded collet type collar holds all wire gauge.