MAC X-Ray Microanalysis Individual Standards, Micro-Analysis Consultants
Supplier: Electron Microscopy Sciences
80074-ZR
80074-IR
80074-OS
80074-AS
80074-C
80074-CO
80074-MO
80074-PD
80074-RH
80074-TL
80074-Y
80074-RU
80074-SC
80074-TE
80074-TI
80074-ZN
80074-AG
80074-CD
80074-NB
80074-V
80074-W
80074-AU
80074-CU
80074-FE
80074-TA
80074-CR
80074-HF
80074-MG
80074-MN
80074-PT
80074-BAS
80074-BAS-FC
80074-BAS-S
80074-NBS
80074-NBS-CA
80074-NBS-HS
80074-NBS-SL
80074-NBS-SS
80074-NI
100495-134EA
927.52
CAD
100495-134
100495-076
100495-096
100495-040
100495-058
100495-062
100495-082
100495-098
100495-104
100495-124
100495-130
100495-106
100495-110
100495-120
100495-122
100495-132
100495-036
100495-060
100495-084
100495-126
100495-128
100495-042
100495-066
100495-068
100495-118
100495-064
100495-074
100495-078
100495-080
100495-100
102099-010
102099-012
102099-014
102099-018
102099-020
102099-022
102099-024
102099-026
102099-028
MAC X-Ray Microanalysis Individual Standards, Micro-Analysis Consultants
Standards
Colour Reference and Spectroscopy Standards
All MAC X-ray Microanalysis standards are suitable for electron-beam energy dispersive or wavelength dispersive X-ray microanalysis systems.
All the standards are polished to a 0.25 micron diamond finish and carbon coated. Supplied in 2, 3, and 5 mm diameter and a location map for standard identification. They are supplied with fully authenticated certificates of analysis.