Horizontal and Vertical Scale Graticules, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
68010-21 67987-19 67992-16 67993-21 67988-19 68006-19 68006-21 67993-16 67993-19 67988-21 67992-21 67987-21 67988-16 67976-21 67976-50 67977-16 67977-21 67977-50 68010-20.4 68010-23 68010-24.5 68010-25 68010-26 68010-27
100499-138EA 298.78 CAD
100499-138 100499-040 100499-062 100499-072 100499-046 100499-118 100499-120 100499-068 100499-070 100499-048 100499-066 100499-042 100499-044 102096-008 102096-010 102096-012 102096-016 102096-018 103303-590 103303-592 103303-594 103303-596 103303-598 103303-600
Horizontal and Vertical Scale Graticules, Electron Microscopy Sciences
Graticules, Reticles and Grids
These scales are used for the measuring of lengths of specimen or distances between points on a variety of different shaped objects.
NE1 scale showed: This eyepiece graticule has an overall length of 10.00 mm with 100 subdivisions of 0.1 mm. When used with a 10x objective each division will represent 10 microns on the specimen. By dividing the division of the chosen graticule by the magnification of the objective one obtains an approximate value that each division will represent on the stage.
Ordering information: Scales in mm and inches available.