Histology Containers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64233-14 64233-26 64233-11 64233-16 64233-17 64233-20 64233-22 64233-12 64233-24
100490-444EA 111.82 CAD
100490-444 100490-458 100490-438 100490-440 100490-446 100490-448 100490-452 100490-450 100490-454 100490-442 100490-456
Histology Containers, Electron Microscopy Sciences
Sample Containers General Purpose Containers
The 20 m to 1 L sizes are supplied with tight-fitting screw caps and warning labels for 10% Formalin with extra space for writing
  • Designed for storage and transport of histology specimens
  • Made from durable polypropylene (except 8L size which is made from polyethylene).
  • Leak proof seal caps.
The 2 to 8L sizes are supplied with snap-top lids and carrying handles, and has no graduations.
All of the containers (except the 5 mL size) are graduated on three sides.