High Precision Diamond Scribing Tools, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
62105-S1 62105-HC 62105-00 62105-B1 62105-BE 62105-B2 62105-S2 62105-HS
100497-830EA 162.04 CAD
100497-830 100501-684 100497-822 100497-824 100501-682 100497-826 100497-832 100497-828
High Precision Diamond Scribing Tools, Electron Microscopy Sciences
Pens Stylus and Scribers
A newly designed diamond scribing tool set containing eight precision natural diamond scribes in various sizes and configurations. This set is perfect for precision work in microscopy. The set consists of eight diamond tips, 3 mm in diameter and are mounted in interchangeable tool cones and an MT-1 tool handle. The tool and handle are all housed in a hardwood instrument case. Individual scribers can be purchased separately.
Order Now
SPECIFICATIONS
Environmentally Preferable