Veco Hexagonal Mesh Grids, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
H100-CU H111K-CU H075-CU H111K-NI H200-CU H150-CU H150-NI H200-AU H400-CU H100-NI H075-NI H200-NI
100495-794EA 79.54 CAD
100495-794 100502-346 100495-790 100495-798 100495-812 100495-802 100495-804 100495-810 100495-822 100495-796 100495-792 100495-814
Veco Hexagonal Mesh Grids, Electron Microscopy Sciences
Graticules, Reticles and Grids
Veco Grids offer superior handling characteristics
Veco Grids are the most rigid grids available. Measures 3.05 mm in diameter. Material: Copper (Cu); Nickel (Ni); Gold (Au).
Plus, With a 0.8 mil thickness.