Gilder Hexagonal Mesh Thin Bar Grids, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
T300H-CU T600H-CU T200H-CU T300H-NI T400H-CU T400H-NI T600H-NI
100496-046EA 157.81 CAD
100496-046 100496-058 100496-036 100496-048 100502-362 100496-054 100496-060
Gilder Hexagonal Mesh Thin Bar Grids, Electron Microscopy Sciences
Graticules, Reticles and Grids
Thin Bar Grids have been developed using a new technology to produce ultra-fine grids with thinner cross bars than regular grids. The result is equally firm specimen support but with 40% more open area for viewing maximum specimen surface area.
  • Made of copper (Cu) or Nicker (Ni)
  • With 3.05" diameter