Headband with Magnifier, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
68108-04 68108-07 68108-15 68108-01 68108-13 68108-16 68108-06 68108-11 68108-12 68108-14 68108-02 68108-17 68108-03
100492-690EA 110.52 CAD
100492-690 100492-696 100492-706 100501-392 100492-702 100492-708 100492-694 100492-698 100492-700 100492-704 100501-394 100492-710 100501-396
Headband with Magnifier, Electron Microscopy Sciences
Magnifiers
Close-up magnification while leaving both hands free for working.
  • Lightweight, adjustable headband for comfort
  • Tilt up and away when not in use
  • Single lens with no center post means clear, comfortable, unobstructed vision with less eye strain and fatigue
  • Available in a variety of magnifications
The headbands that have two ranges and two magnifications include 3.5X magnification Auxiliary Lens.
Order Now
SPECIFICATIONS
Environmentally Preferable