Veco Handle Grids Hexagonal Mesh, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
HD400H-CU HD400H-NI HD150H-CU HD200H-CU HD300H-CU HD100H-NI HD150H-NI HD300H-NI
100495-866EA 82.35 CAD
100495-866 100495-868 100495-844 100495-850 100495-860 100495-836 100495-846 100495-862
Veco Handle Grids Hexagonal Mesh, Electron Microscopy Sciences
Graticules, Reticles and Grids
Veco Grids offer superior handling characteristics
Veco Grids are the most rigid grids available. Measures 3.05 mm in diameter. Material: Copper (Cu); Nickel (Ni).
Plus, with a 0.8 mil thickness.