Grided Microscope Slides without hydrophobic coating, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
63408-01 63406-01 63406-02 63405-01 63405-02 63409-02
100490-308EA 295.92 CAD
100490-308 100488-872 100490-302 100502-014 100490-300 100490-310
Grided Microscope Slides without hydrophobic coating, Electron Microscopy Sciences
Slides Counting Chambers
These 3×1" slides have an indexed grid on the reverse side of the frosted end. The ink is resistant to stains and common laboratory solvents.
  • Each square grid fills one low-powered-field (approximately 100× magnification)
  • Easy to locate an organism or cell every time
  • Eighty one squares
The slides are available with coatings of Poly-L-Lysine or Silane, or they can be purchased plain (uncoated). The grid measures 2 mm square, 16 mm total. Numbers 1-8 and alpha A-H.