Grid Storage Box, Plastic, 100 Capacity, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71156 71155
100499-706DZ 415.9 CAD
100499-706 100491-748
Grid Storage Box, Plastic, 100 Capacity, Electron Microscopy Sciences
Boxes Storage Boxes
A newly designed grid storage box similar to the original LKB box. Made from a special plastic that minimizes static. Complete with grid recording card.
  • Made from a special plastic that minimizes static
  • Complete with grid recording card