Gloves, Latex, Surgeons, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71114-01 71114-20 71115-01 71115-20 71116-01 71116-20
102097-388EA 40.13 CAD
102097-388 102097-390 102097-394 102097-396 102097-398 102097-402 102097-404 102097-406 102097-410
Gloves, Latex, Surgeons, Electron Microscopy Sciences
Gloves
Made from 100% natural latex with the addition of Aloe Vera and Vitamin E, offering the softness, tactile sensitivity and skin protection.
  • Economical
  • Ambidextrous style
  • Beaded cuff
Chemically resistant to acids, alcohols, ketones and alkalis. Non-sterile.
Available either with powdered (biologically cornstarch dusting, ~2 mg/glove) or powder-free gloves.
Packaging: Packed in a convenient dispenser box, 100/box.