Glass Microbeakers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
60980 60986 60982
100503-374BX 662.51 CAD
100503-374 100503-380 100503-376
Glass Microbeakers, Electron Microscopy Sciences
Beakers
Used for the processing of micro-samples and for visual or microscopic examination, staining, fixation, infiltration, liquid evaporation, bacterial growth, weighing, etc.
Order Now
SPECIFICATIONS
Environmentally Preferable