Gelatin Capsule Holders, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70120-06 70120-04 70120-01
100490-728EA 297.31 CAD
100490-728 100499-566 100502-818
Gelatin Capsule Holders, Electron Microscopy Sciences
Embedding Cassettes Tissue Capsules
The whole capsule is exposed to allow for even heat and/or UV light distribution during polymerization.
  • Made from durable polyethylene
  • It accommodates 15 gelatin capsules size "00"
Measures: 127×76 mm (5×3").