Formvar Carbon Support Film on Hexagonal Grids, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
FCF100H-CU-25 FCF100H-CU-50 FCF200H-CU-25 FCF200H-CU-50 FCF200H-NI-50 FCF300H-CU-25 FCF300H-CU-50 FCF300H-NI-25 FCF400H-CU-25 FCF400H-CU-50 FCF400H-NI-25 FCF300H-CU-SC FCF300H-CU-UB FCF100H-CU-SB FCF100H-NI-TB
102100-470BX 133.83 CAD
102100-470 102100-472 102100-506 102100-508 102100-512 102100-530 102100-532 102100-534 102100-542 102100-544 102100-546 103302-920 103302-924 103303-258 103303-378
Formvar Carbon Support Film on Hexagonal Grids, Electron Microscopy Sciences
Graticules, Reticles and Grids
The formvar coated grids are stabilized with an evaporated carbon film. This type of coating is excellent for specimen support, especially for ultra thin sections. All film is laid on the shiny side of the grid.
  • Formvar coatings thickness of standard (10 nm), ultra-thin (5 to 6 nm), thick (15 to 20 nm), or extra thick (25 to 50 nm)
  • Select copper, nickel, or gold grids in 100, 150, 200, 300 or 400 mesh
  • Carbon coatings thickness of 1 nm, 3 to 4 nm, and 20 to 30 nm
  • Optically checked followed by batch testing in the EM
Packaging: Packed in grid storage box.