Foreline Traps, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
91050-15 91050-25 91050-20
100502-978EA 914.41 CAD
100502-978 100502-328 100502-326
Foreline Traps, Electron Microscopy Sciences
Vacuum Traps
These rechargeable in-line traps effectively block oil backstreaming from mechanical pumps. Any mechanically pumped system, even one with a turbo pump, should have a foreline traps. The trap is placed in the foreline between the roughing pump and the diffusion or turbo pump. It uses a disposable, oxygen-free copper maze absorbent which requires no liquid nitrogen and will not hold water. Typically the maze is replaced every 1-2 years. The seamless body is available in aluminum. Replacement elements (Copper Maze) also available.
  • A quick change clamp for replacement of maze
  • A viton “O”
More Product Information