Foam Tipped Applicator; Foam Swabs, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
72317 72314 72325 72326 72312 72315
100488-472EA 34.73 CAD
100488-472 100488-468 100501-828 100501-830 100488-466 100488-470
Foam Tipped Applicator; Foam Swabs, Electron Microscopy Sciences
Swabs Swabs for Cleaning
The 6" Wooden Shaft Applicator is round, foam-tip over regular-cotton tip on a 6" long wooden shaft
These foam tipped applicators are lint free, non-abrasive, and sized for precision cleaning with solvents. (Freon, Trichloroethane, Isopropyl Alcohol, and Acetone). The head is made from 100% urethane foam with 100 PPI porosity. Tips are shred resistant and bonded securely to the shaft.
The 6" Wooden Shaft Applicator is round, foam-tip over regular-cotton tip on a 6" long wooden shaft. Head size d x L: 3/8 x 1 3/16". Packed 50 sticks in a sealed bag.
The 5" Shaft, Rectangular Foam Tip is polypropylene shaft. Head size d x L: 1 1/16 x 1". Packed 50 sticks in a sealed bag.
The 6" Wooden Shaft Anti-Static Foam-Tip Over Cotton Tip measures d x L: 3/8 x 1 3/16". Packed 50 sticks in a sealed bag.
Head size d x L: 3/8 x 1 3/16". Packed 50 sticks in a sealed bag.