Flexible Shaft Machine - General Application Kit, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
62134-20 62133-10 62133-20 62134-10 62130-20 62131-10 62131-20 62130-10
100497-898EA 436.9 CAD
100497-898 100497-892 100497-894 100497-896 100497-880 100497-884 100497-886 100497-878
Flexible Shaft Machine - General Application Kit, Electron Microscopy Sciences
Tool Sets
A versatile hang-up flexible shaft machine.
Accessories information: Mounted accessories are on 2.35 mm (3/32") arbors.
Ordering information: Available either 1/10 or 1/8 HP ball bearing motor, variable speed 0 to 18 rpm. Complete kit comes with a hand piece, 1 m (39") Flexible shaft, variable speed foot pedal, and a bracket for hanging. Lifetime warranty on motor under normal use.