Flat Emery Sticks, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
62119-04 62119-20 62119-10 62119-30 62119-02
100499-880DZ 53.37 CAD
100499-880 100499-884 100499-882 100499-886 100499-876
Flat Emery Sticks, Electron Microscopy Sciences
Abrasives
Emery paper which is glued to a wooden handle. Very useful for filing excess resin of sample block tips. Grit size ranges from number 4/0 (finest) to 3 (coarsest).
Dimensions L x W: 279 x 19.1 mm (11 x 3/4").
Order Now
SPECIFICATIONS
Environmentally Preferable