Finger Cots, Latex, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71132 71131 71130
100488-428EA 25.98 CAD
100488-428 100488-426 100488-424
Finger Cots, Latex, Electron Microscopy Sciences
Finger Cots Latex Finger Cots
Inexpensive, expandable finger cots.
  • Seamless
  • Protective dexterity without covering the entire hand
  • Powderless
  • Non-rolled
3 1/2 mil (0.0030"). Length 40 mm (2 3/4").